Disodium-4-amino-4'-nitrostilbene-2,2'-sulfonate ≥80.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-Amino-4'-nitrostilbene-2,2'-disulfonic Acid Disodium Salt
, 4-Amino-4'-nitrostilbene-2,2'-disulphonic Acid Disodium Salt
A1199-500G
A1199-25G
TCA1199-500GEA
695
USD
TCA1199-500G
TCA1199-025G
Disodium-4-amino-4'-nitrostilbene-2,2'-sulfonate ≥80.0% (by titrimetric analysis)
Disodium-4-amino-4'-nitrostilbene-2,2'-sulfonate
CAS Number: 6634-82-8
MDL Number: MFCD00143251
Molecular Formula: C14H12N2O8S2
Molecular Weight: 444.34
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 4-Amino-4'-nitrostilbene-2,2'-disulfonic Acid Disodium Salt
Formula:
C₁₄H₁₂N₂O₈S₂ MW: 444.34 g/mol |
MDL Number:
MFCD00143251 CAS Number: 6634-82-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...