Dihexyl malonate ≥98.0%

Supplier: TCI America

Synonyms: Malonic Acid Dihexyl Ester

D3164-5G
TCD3164-5GEA 123.22 USD
TCD3164-5G
Dihexyl malonate ≥98.0%
Dihexyl malonate

CAS Number: 1431-37-4
MDL Number: MFCD06797099
Molecular Formula: C15H28O4
Molecular Weight: 272.39
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Malonic Acid Dihexyl Ester

Formula: C₁₅H₂₈O₄
MW: 272.39 g/mol
Density: 0.95 (20 °C)
MDL Number: MFCD06797099
CAS Number: 1431-37-4
https://us.vwr.com/stibo/web/std.lang.all/01/67/28980167.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR