Di-n-octyl sulfone ≥98.0%

Supplier: TCI America

Synonyms: n-Octyl Sulfonen-Octyl Sulphone

D3161-5G
TCD3161-5GEA 88.26 USD
TCD3161-5G
Di-n-octyl sulfone ≥98.0%
Di-n-octyl sulfone

CAS Number: 7726-20-7
MDL Number: MFCD00039760
Molecular Formula: C16H34O2S
Molecular Weight: 290.51
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 78


Product Categories: Synthetic Organic Chemistry, Sulfones
Other Product Names / Synonyms: n-Octyl Sulfone

Formula: C₁₆H₃₄O₂S
MW: 290.51 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00039760
CAS Number: 7726-20-7
https://us.vwr.com/stibo/web/std.lang.all/01/65/28980165.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR