3',4'-Dimethylbenzophenone-2-carboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D2978-5G
TCD2978-5GEA
184.65
USD
TCD2978-5G
3',4'-Dimethylbenzophenone-2-carboxylic acid ≥98.0% (by titrimetric analysis)
3',4'-Dimethylbenzophenone-2-carboxylic acid
CAS Number: 2159-42-4
Molecular Formula: C16H14O3
Molecular Weight: 254.29
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 168
Other Product Names / Synonyms: 2-(3,4-Dimethylbenzoyl)benzoic Acid
Formula:
C₁₆H₁₄O₃ Melting Pt: 168 °C |
CAS Number:
2159-42-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...