(2,4-Dichlorophenyl)acetone ≥98.0%

Supplier: TCI America

Synonyms: 1-(2,4-Dichlorophenyl)-2-propanone2,4-Dichlorophenylacetone

D2928-25G D2928-5G
TCD2928-25GEA 392.43 USD
TCD2928-25G TCD2928-5G
(2,4-Dichlorophenyl)acetone ≥98.0%
(2,4-Dichlorophenyl)acetone

CAS Number: 93457-07-9
MDL Number: MFCD00210405
Molecular Formula: C9H8Cl2O
Molecular Weight: 203.06
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 49


Other Product Names / Synonyms: 1-(2,4-Dichlorophenyl)-2-propanone

https://us.vwr.com/stibo/web/std.lang.all/00/75/28980075.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR