2,4-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2925-5G
TCD2925-5GEA 62.9 USD
TCD2925-5G
2,4-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)
2,4-Dichlorophenethylamine

CAS Number: 52516-13-9
MDL Number: MFCD00060611
Molecular Formula: C8H9Cl2N
Molecular Weight: 190.07
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.27


Other Product Names / Synonyms: 1-Amino-2-(2,4-dichlorophenyl)ethane, 1-(2-Aminoethyl)-2,4-dichlorobenzene, 2,4-Dichlorophenethylamine

https://us.vwr.com/stibo/web/std.lang.all/00/72/28980072.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR