(4S,5S)-1,3-Dimethyl-4,5-diphenyl-2-[(R)-1-benzyl-2-hydroxyethylimino]imidazolidine ≥98.0% (by HPLC, total nitrogen)

D2899-100MG
TCD2899-100MGEA 259.08 USD
TCD2899-100MG
(4S,5S)-1,3-Dimethyl-4,5-diphenyl-2-[(R)-1-benzyl-2-hydroxyethylimino]imidazolidine ≥98.0% (by HPLC, total nitrogen)
(4S,5S)-1,3-Dimethyl-4,5-diphenyl-2-[(R)-1-benzyl-2-hydroxyethylimino]imidazolidine

CAS Number: 210468-90-9
MDL Number: MFCD06797086
Molecular Formula: C26H29N3O
Molecular Weight: 399.54
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 114
Specific rotation [a]20/D: 75 deg (C=1, CHCl3)

Formula: C₂₆H₂₉N₃O
Melting Pt: 114 °C
MDL Number: MFCD06797086
CAS Number: 210468-90-9
https://us.vwr.com/stibo/web/std.lang.all/00/44/28980044.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR