1,3-Dichloro-2,4,6-trifluorobenzene ≥97.0%

Supplier: TCI America

Synonyms: 2,4-Dichloro-1,3,5-trifluorobenzene

D2868-5G
TCD2868-5GEA 257.7 USD
TCD2868-5G
1,3-Dichloro-2,4,6-trifluorobenzene ≥97.0%
1,3-Dichloro-2,4,6-trifluorobenzene

CAS Number: 2368-53-8
MDL Number: MFCD00012242
Molecular Formula: C6HCl2F3
Molecular Weight: 200.97
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.60

Formula: C₆HCl₂F₃
MW: 200.97 g/mol
Density: 1.60 (20 °C)
MDL Number: MFCD00012242
CAS Number: 2368-53-8
https://us.vwr.com/stibo/web/std.lang.all/00/23/28980023.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR