8-Dimethylamino-1-octanol ≥93.0%
Supplier: TCI America
D2846-5G
D2846-1G
TCD2846-1GEA
112.61
USD
TCD2846-1G
TCD2846-5G
8-Dimethylamino-1-octanol ≥93.0%
8-Dimethylamino-1-octanol
CAS Number: 29823-87-8
MDL Number: MFCD03701100
Molecular Formula: C10H23NO
Molecular Weight: 173.30
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 0.86
Formula:
C₁₀H₂₃NO Density: 0.86 (20 °C) |
MDL Number:
MFCD03701100 CAS Number: 29823-87-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...