(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0%
Supplier: TCI America
Synonyms:
(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene
D2802-200MG
TCD2802-200MGEA
179
USD
TCD2802-200MG
(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0%
(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl
CAS Number: 75714-60-2
MDL Number: MFCD04038415
Molecular Formula: C22H16Br2O2
Molecular Weight: 472.18
Purity/Analysis Method:
Form: Crystal
Formula:
C₂₂H₁₆Br₂O₂ MW: 472.18 g/mol |
MDL Number:
MFCD04038415 CAS Number: 75714-60-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...