Ethyl-3-(dimethylamino)-3-ethoxy-2,2-difluoropropionate ≥95.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 3-(Dimethylamino)-3-ethoxy-2,2-difluoropropionic Acid Ethyl Ester

D2782-5G D2782-1G
TCD2782-5GEA 550 USD
TCD2782-5G TCD2782-1G
Ethyl-3-(dimethylamino)-3-ethoxy-2,2-difluoropropionate ≥95.0% (by total nitrogen basis)
Ethyl-3-(dimethylamino)-3-ethoxy-2,2-difluoropropionate

CAS Number: 141546-96-5
MDL Number: MFCD00722253
Molecular Formula: C9H17F2NO3
Molecular Weight: 225.24
Purity/Analysis Method: 95.0% (N)
Form: Clear Liquid
Boiling point (°C): 62
Specific Gravity (20/20): 1.09
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Dimethylamino)-3-ethoxy-2,2-difluoropropionic Acid Ethyl Ester

Formula: C₉H₁₇F₂NO₃
MW: 225.24 g/mol
Boiling Pt: 62 °C
Density: 1.09 (20 °C)
MDL Number: MFCD00722253
CAS Number: 141546-96-5
https://us.vwr.com/stibo/web/std.lang.all/99/26/28979926.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR