Diundecylamine ≥97.0%

Supplier: TCI America

D2769-5G
TCD2769-5GEA 479.96 USD
TCD2769-5G
Diundecylamine ≥97.0%
Diundecylamine

CAS Number: 16165-33-6
MDL Number: MFCD00026548
Molecular Formula: C22H47N
Molecular Weight: 325.63
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 232
Melting point (°C): 49

Formula: C₂₂H₄₇N
Boiling Pt: 232 °C
Melting Pt: 49 °C
MDL Number: MFCD00026548
CAS Number: 16165-33-6
https://us.vwr.com/stibo/web/std.lang.all/99/12/28979912.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR