Dimethyldioctylammonium bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D2754-5G D2754-25G
TCD2754-5GEA 165.4 USD
TCD2754-5G TCD2754-25G
Dimethyldioctylammonium bromide ≥97.0% (by titrimetric analysis)
Dimethyldioctylammonium bromide

CAS Number: 3026-69-5
MDL Number: MFCD00082931
Molecular Formula: C18H40BrN
Molecular Weight: 350.43
Purity/Analysis Method: 97.0% (T)
Form: Crystal

Formula: C₁₈H₄₀BrN
MDL Number: MFCD00082931
CAS Number: 3026-69-5
https://us.vwr.com/stibo/web/std.lang.all/98/89/28979889.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR