Methyl-3,5-di-tert-butylbenzoate ≥98.0%
Supplier: TCI America
Synonyms:
3,5-Di-tert-butylbenzoic Acid Methyl Ester
D2695-5G
TCD2695-005GEA
320.65
USD
TCD2695-005G
Methyl-3,5-di-tert-butylbenzoate ≥98.0%
Methyl-3,5-di-tert-butylbenzoate
CAS Number: 64277-87-8
MDL Number: MFCD03844786
Molecular Formula: C16H24O2
Molecular Weight: 248.37
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 53
Other Product Names / Synonyms: 3,5-Di-tert-butylbenzoic Acid Methyl Ester
Formula:
C₁₆H₂₄O₂ MW: 248.37 g/mol Melting Pt: 53 °C |
MDL Number:
MFCD03844786 CAS Number: 64277-87-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...