Diheptylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2675-5G
TCD2675-5GEA 324 USD
TCD2675-5G
Diheptylamine ≥98.0% (by GC, titration analysis)
Diheptylamine

CAS Number: 2470-68-0
MDL Number: MFCD00027309
Molecular Formula: C14H31N
Molecular Weight: 213.41
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 148
Specific Gravity (20/20): 0.79

https://us.vwr.com/stibo/web/std.lang.all/96/88/28979688.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR