TCIAA1149

A1149-25ML
TCA1149-025MLEA 40.07 USD
TCA1149-025ML
TCIAA1149
CAS Number: 40853-55-2
MDL Number: MFCD00129077
Molecular Formula: C12H24O2
Molecular Weight: 200.32
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Specific Gravity (20/20): 0.87

Other Product Names / Synonyms: Acetic Acid 2-Isopropyl-5-methylhexyl Ester, Tetrahydrolavandulyl Acetate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR