2,4'-Dihydroxybenzophenone ≥98.0%

Supplier: TCI America

D2664-5G
TCD2664-5GEA 260 USD
TCD2664-5G
2,4'-Dihydroxybenzophenone ≥98.0%
2,4'-Dihydroxybenzophenone

CAS Number: 606-12-2
MDL Number: MFCD02181674
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 148

Formula: C₁₃H₁₀O₃
Melting Pt: 148 °C
MDL Number: MFCD02181674
CAS Number: 606-12-2
https://us.vwr.com/stibo/web/std.lang.all/96/52/28979652.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR