Diethyl(3-pyridyl)borane ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3-(Diethylboryl)pyridine

D2661-1G
TCD2661-001GEA 19 USD
TCD2661-001G
Diethyl(3-pyridyl)borane ≥98.0% (by HPLC)
Diethyl(3-pyridyl)borane

CAS Number: 89878-14-8
MDL Number: MFCD00012348
Molecular Formula: C9H14BN
Molecular Weight: 147.03
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 162


Other Product Names / Synonyms: 3-(Diethylboryl)pyridine

Formula: C₉H₁₄BN
MW: 147.03 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00012348
CAS Number: 89878-14-8
https://us.vwr.com/stibo/web/std.lang.all/96/42/28979642.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR