trans-2-Heptenyl acetate ≥97.0%
Supplier: TCI America
A1145-10ML
TCA1145-010MLEA
121
USD
TCA1145-010ML
trans-2-Heptenyl acetate ≥97.0%
trans-2-Heptenyl acetate
CAS Number: 16939-73-4
MDL Number: MFCD00036584
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 193
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: Acetic Acid trans-2-Heptenyl Ester
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...