1,2:3,5-Di-O-isopropylidene-ɑ-D-xylofuranose ≥98.0%

Supplier: TCI America

Synonyms: (3aR,3bS,7aR,8aR)-2,2,5,5-Tetramethyltetrahydro-7H-[1,3]dioxolo[4',5':4,5]furo[3,2-d][1,3]dioxine

D2616-5G
TCD2616-005GEA 114.65 USD
TCD2616-005G
1,2:3,5-Di-O-isopropylidene-ɑ-D-xylofuranose ≥98.0%
1,2:3,5-Di-O-isopropylidene-alpha-D-xylofuranose

CAS Number: 20881-04-3
MDL Number: MFCD00063224
Molecular Formula: C11H18O5
Molecular Weight: 230.26
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 45
Specific rotation [a]20/D: 19 deg (C=3, Acetone)

Formula: C₁₁H₁₈O₅
MW: 230.26 g/mol
Boiling Pt: 125 °C
Melting Pt: 45 °C
MDL Number: MFCD00063224
CAS Number: 20881-04-3
https://us.vwr.com/stibo/web/std.lang.all/95/86/28979586.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR