Ethyl-2,3-dibromobutyrate ≥95.0%

Supplier: TCI America

Synonyms: 2,3-Dibromobutyric Acid Ethyl EsterEthyl 2,3-dibromobutanoate

D2612-5G
TCD2612-005GEA 221.18 USD
TCD2612-005G
Ethyl-2,3-dibromobutyrate ≥95.0%
Ethyl-2,3-dibromobutyrate

CAS Number: 609-11-0
MDL Number: MFCD00017863
Molecular Formula: C6H10Br2O2
Molecular Weight: 273.95
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Specific Gravity (20/20): 1.69


Other Product Names / Synonyms: 2,3-Dibromobutyric Acid Ethyl Ester

Formula: C₆H₁₀Br₂O₂
MW: 273.95 g/mol
Boiling Pt: 104 °C
Density: 1.69 (20 °C)
MDL Number: MFCD00017863
CAS Number: 609-11-0
https://us.vwr.com/stibo/web/std.lang.all/95/82/28979582.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR