Dichlorodiisopropylsilane ≥98.0%

Supplier: TCI America

Synonyms: Diisopropyldichlorosilane

D2603-5G D2603-25G
TCD2603-025GEA 125.83 USD
TCD2603-025G TCD2603-005G
Dichlorodiisopropylsilane ≥98.0%
Dichlorodiisopropylsilane

CAS Number: 7751-38-4
MDL Number: MFCD00054895
Molecular Formula: C6H14Cl2Si
Molecular Weight: 185.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 43
Specific Gravity (20/20): 1.03

Formula: C₆H₁₄Cl₂Si
MW: 185.16 g/mol
Boiling Pt: 170 °C
Density: 1.03 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00054895
CAS Number: 7751-38-4
https://us.vwr.com/stibo/web/std.lang.all/95/73/28979573.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR