6,6'-Dithiodinicotinic acid ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 6,6'-Dithiobis(3-pyridinecarboxylic Acid)

D2564-25G D2564-5G
TCD2564-25GEA 435.34 USD
TCD2564-25G TCD2564-005G
6,6'-Dithiodinicotinic acid ≥98.0% (by HPLC)
6,6'-Dithiodinicotinic acid

CAS Number: 15658-35-2
MDL Number: MFCD00006454
Molecular Formula: C12H8N2O4S2
Molecular Weight: 308.33
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 6,6'-Dithiobis(3-pyridinecarboxylic Acid)

Formula: C₁₂H₈N₂O₄S₂
MW: 308.33 g/mol
MDL Number: MFCD00006454
CAS Number: 15658-35-2
https://us.vwr.com/stibo/web/std.lang.all/95/37/28979537.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR