1,3-Dimethyl-2-thiohydantoin ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dimethyl-2-thioxoimidazolidin-4-one

D2506-5G
TCD2506-005GEA 200 USD
TCD2506-005G
1,3-Dimethyl-2-thiohydantoin ≥98.0%
1,3-Dimethyl-2-thiohydantoin

CAS Number: 1801-62-3
MDL Number: MFCD00186242
Molecular Formula: C5H8N2OS
Molecular Weight: 144.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 93

Formula: C₅H₈N₂OS
MW: 144.19 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00186242
CAS Number: 1801-62-3
https://us.vwr.com/stibo/web/std.lang.all/94/34/28979434.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR