Diethyl dodecanedioate ≥97.0%
Supplier: TCI America
Synonyms:
Dodecanedioic Acid Diethyl Ester
D2486-25G
TCD2486-025GEA
132.9
USD
TCD2486-025G
Diethyl dodecanedioate ≥97.0%
Diethyl dodecanedioate
CAS Number: 10471-28-0
MDL Number: MFCD00009204
Molecular Formula: C16H30O4
Molecular Weight: 286.41
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: Dodecanedioic Acid Diethyl Ester
Formula:
C₁₆H₃₀O₄ MW: 286.41 g/mol Density: 0.95 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00009204 CAS Number: 10471-28-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...