Dibutyltin bis(trifluoromethanesulfonate) ≥98.0% (by gravimetric analysis)
Supplier: TCI America
Synonyms:
Dibutyltin Ditriflate
D2466-5G
TCD2466-005GEA
187.7
USD
TCD2466-005G
Dibutyltin bis(trifluoromethanesulfonate) ≥98.0% (by gravimetric analysis)
Dibutyltin bis(trifluoromethanesulfonate)
CAS Number: 38438-11-8
MDL Number: MFCD00671529
Molecular Formula: C10H18F6O6S2Sn
Molecular Weight: 531.07
Purity/Analysis Method:
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Homogeneous Catalysts, Homogeneous Catalysts
Other Product Names / Synonyms: Dibutyltin Ditriflate
Formula:
C₁₀H₁₈F₆O₆S₂Sn MW: 531.07 g/mol |
MDL Number:
MFCD00671529 CAS Number: 38438-11-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...