Diethyl-2,5-dioxahexanedioate ≥98.0%

Supplier: TCI America

Synonyms: 2,5-Dioxahexanedioic Acid Diethyl Ester

D2456-25G
TCD2456-025GEA 257.7 USD
TCD2456-025G
Diethyl-2,5-dioxahexanedioate ≥98.0%
Diethyl-2,5-dioxahexanedioate

CAS Number: 35466-87-6
MDL Number: MFCD00460005
Molecular Formula: C8H14O6
Molecular Weight: 206.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 227
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 2,5-Dioxahexanedioic Acid Diethyl Ester

Formula: C₈H₁₄O₆
MW: 206.19 g/mol
Boiling Pt: 227 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00460005
CAS Number: 35466-87-6
https://us.vwr.com/stibo/web/std.lang.all/93/78/28979378.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR