Acetone dimethylhydrazone ≥98.0%

Supplier: TCI America

A1116-25ML
TCA1116-025MLEA 163 USD
TCA1116-025ML
Acetone dimethylhydrazone ≥98.0%
Acetone dimethylhydrazone

CAS Number: 13483-31-3
MDL Number: MFCD00059022
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Specific Gravity (20/20): 0.80

Formula: C₅H₁₂N₂
Boiling Pt: 95 °C
Density: 0.80 (20 °C)
MDL Number: MFCD00059022
CAS Number: 13483-31-3
https://us.vwr.com/stibo/web/std.lang.all/89/35/28928935.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR