16-Doxylstearic acid ≥97.0% (by HPLC, titration analysis) free radical
Synonyms:
2-(14-Carboxytetradecyl)-2-ethyl-4,4-dimethyl-3-oxazolidinyloxy
D2399-100MG
D2399-25MG
TCD2399-100MGEA
634.48
USD
TCD2399-100MG
TCD2399-25MG
16-Doxylstearic acid ≥97.0% (by HPLC, titration analysis) free radical
16-Doxylstearic acid
CAS Number: 53034-38-1
MDL Number: MFCD00010066
Molecular Formula: C22H42NO4
Molecular Weight: 384.58
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 58
Flash Point (°C): 110
Storage Temperature:
Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Analytical Science, Spin Labels
Other Product Names / Synonyms: 2-(14-Carboxytetradecyl)-2-ethyl-4,4-dimethyl-3-oxazolidinyloxy
Formula:
C₂₂H₄₂NO₄ MW: 384.58 g/mol Melting Pt: 58 °C Flash Pt: 110 °C |
MDL Number:
MFCD00010066 CAS Number: 53034-38-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...