Methyl-2,2-dimethoxypropionate ≥96.0%

Supplier: TCI America

Synonyms: Methyl 2,2-dimethoxypropanoate

D2382-25G
TCD2382-025GEA 271.9 USD
TCD2382-025G
Methyl-2,2-dimethoxypropionate ≥96.0%
Methyl 2,2-dimethoxypropionate

CAS Number: 10076-48-9
MDL Number: MFCD00048066
Molecular Formula: C6H12O4
Molecular Weight: 148.16
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 2,2-Dimethoxypropionic Acid Methyl Ester

https://us.vwr.com/stibo/web/std.lang.all/93/07/28979307.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR