Dichlorododecylmethylsilane ≥95.0%

Supplier: TCI America

Synonyms: Dichloro(dodecyl)(methyl)silane

D2349-25ML
TCD2349-025MLEA 182 USD
TCD2349-025ML
Dichlorododecylmethylsilane ≥95.0%
Dichlorododecylmethylsilane

CAS Number: 18407-07-3
MDL Number: MFCD00018082
Molecular Formula: C13H28Cl2Si
Molecular Weight: 283.35
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 143
Specific Gravity (20/20): 0.95

Formula: C₁₃H₂₈Cl₂Si
MW: 283.35 g/mol
Boiling Pt: 127 °C
Density: 0,95 (20 °C)
Flash Pt: 143 °C
MDL Number: MFCD00018082
CAS Number: 18407-07-3

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR