1,4-Dihydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1,4-Dihydroxy-2-naphthalenecarboxylic acid

D2296-25G
TCD2296-025GEA 56.84 USD
TCD2296-025G
1,4-Dihydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
1,4-Dihydroxy-2-naphthoic acid

CAS Number: 31519-22-9
MDL Number: MFCD00010370
Molecular Formula: C11H8O4
Molecular Weight: 204.18
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 191


Other Product Names / Synonyms: 1,4-Dihydroxy-2-naphthalenecarboxylic Acid

Formula: C₁₁H₈O₄
MW: 204.18
Melting Pt: 191 °C
MDL Number: MFCD00010370
CAS Number: 31519-22-9
https://us.vwr.com/stibo/web/std.lang.all/92/27/28979227.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR