1,3-Dimethyl-2-(2-thienyl)imidazolidine

Supplier: TCI America

D2289-5G
TCD2289-005GEA 187 USD
TCD2289-005G
1,3-Dimethyl-2-(2-thienyl)imidazolidine
1,3-Dimethyl-2-(2-thienyl)imidazolidine

CAS Number: 104208-13-1
MDL Number: MFCD00191636
Molecular Formula: C9H14N2S
Molecular Weight: 182.29
Form: Clear Liquid
Color: Slightly Pale Yellow

Formula: C₉H₁₄N₂S
MDL Number: MFCD00191636
CAS Number: 104208-13-1
https://us.vwr.com/stibo/web/std.lang.all/92/20/28979220.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR