6,6'-Dihydroxy-4,4,4',4',7,7'-hexamethyl-2,2'-spirobichroman ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,4,4',4',7,7'-Hexamethyl-2,2'-spirobi[chroman]-6,6'-diol

D2286-25G
TCD2286-025GEA 193.8 USD
TCD2286-025G
6,6'-Dihydroxy-4,4,4',4',7,7'-hexamethyl-2,2'-spirobichroman ≥98.0% (by HPLC)
6,6'-Dihydroxy-4,4,4',4',7,7'-hexamethyl-2,2'-spirobichroman

CAS Number: 40278-59-9
MDL Number: MFCD00191424
Molecular Formula: C23H28O4
Molecular Weight: 368.47
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 210


Other Product Names / Synonyms: 4,4,4',4',7,7'-Hexamethyl-2,2'-spirobi[chroman]-6,6'-diol

Formula: C₂₃H₂₈O₄
MW: 368.47 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00191424
CAS Number: 40278-59-9
https://us.vwr.com/stibo/web/std.lang.all/92/17/28979217.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR