Chlorodiethylisopropylsilane ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Diethylisopropylsilyl Chloride
D2262-1G
D2262-5G
TCD2262-001GEA
91.32
USD
TCD2262-001G
TCD2262-5G
Chlorodiethylisopropylsilane ≥97.0% (by titrimetric analysis)
Chlorodiethylisopropylsilane
CAS Number: 107149-56-4
MDL Number: MFCD00191631
Molecular Formula: C7H17ClSi
Molecular Weight: 164.75
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 38
Specific Gravity (20/20): 0.90
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Diethylisopropylsilyl Chloride
Formula:
C₇H₁₇ClSi MW: 164.75 g/mol Boiling Pt: 75 °C Density: 0.90 (20 °C) Flash Pt: 38 °C |
MDL Number:
MFCD00191631 CAS Number: 107149-56-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...