(+)-1,4-Di-O-benzyl-D-threitol ≥98.0%
Supplier: TCI America
Synonyms:
(+)-(2R,3R)-1,4-Bis(benzyloxy)-2,3-butanediol
D2239-1G
TCD2239-001GEA
454
USD
TCD2239-001G
(+)-1,4-Di-O-benzyl-D-threitol ≥98.0%
(+)-1,4-Di-O-benzyl-D-threitol
CAS Number: 91604-41-0
MDL Number: MFCD00077722
Molecular Formula: C18H22O4
Molecular Weight: 302.37
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 56
Specific rotation [a]20/D: 6.3 deg (C=5, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Sugar Alcohols, Sugars (Others)
Other Product Names / Synonyms: (+)-(2R,3R)-1,4-Bis(benzyloxy)-2,3-butanediol
Formula:
C₁₈H₂₂O₄ MW: 302.37 g/mol Melting Pt: 56 °C |
MDL Number:
MFCD00077722 CAS Number: 91604-41-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...