Dimethylphenacylsulfonium tetrafluoroborate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2231-1G D2231-5G
TCD2231-001GEA 90.29 USD
TCD2231-001G TCD2231-5G
Dimethylphenacylsulfonium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Dimethylphenacylsulphonium tetrafluoroborate

CAS Number: 24806-57-3
MDL Number: MFCD00191352
Molecular Formula: C10H13BF4OS
Molecular Weight: 268.08
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 170

Formula: C₁₀H₁₃BF₄OS
Melting Pt: 170 °C
MDL Number: MFCD00191352
CAS Number: 24806-57-3
https://us.vwr.com/stibo/web/std.lang.all/91/27/28979127.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR