meso-2,3-Diphenylsuccinonitrile ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: alpha,beta-Dicyanobibenzylalpha,beta-Dicyanodibenzyl

D2219-5G
TCD2219-005GEA 143 USD
TCD2219-005G
meso-2,3-Diphenylsuccinonitrile ≥97.0% (by HPLC)
meso-2,3-Diphenylsuccinonitrile

MDL Number: MFCD00019804
Molecular Formula: C16H12N2
Molecular Weight: 232.29
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: alpha,beta-Dicyanobibenzyl, alpha,beta-Dicyanodibenzyl

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR