Dicyclohexylphosphinyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2205-5G
TCD2205-005GEA 215.12 USD
TCD2205-005G
Dicyclohexylphosphinyl chloride ≥98.0% (by GC, titration analysis)
Dicyclohexylphosphinyl chloride

CAS Number: 15873-72-0
MDL Number: MFCD00191344
Molecular Formula: C12H22ClOP
Molecular Weight: 248.73
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 109
Storage Temperature: 0-10°C

Formula: C₁₂H₂₂ClOP
Melting Pt: 109 °C
MDL Number: MFCD00191344
CAS Number: 15873-72-0
https://us.vwr.com/stibo/web/std.lang.all/90/72/28979072.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR