(1-Aminoethyl)phosphonic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

A1071-1G
TCA1071-001GEA 387.56 USD
TCA1071-001G
(1-Aminoethyl)phosphonic acid ≥95.0% (by titrimetric analysis)
(1-Aminoethyl)phosphonic acid

CAS Number: 6323-97-3
MDL Number: MFCD00008068
Molecular Formula: C2H8NO3P
Molecular Weight: 125.06
Purity/Analysis Method: 95.0% (T)
Form: Crystal

Formula: C₂H₈NO₃P
MDL Number: MFCD00008068
CAS Number: 6323-97-3
https://us.vwr.com/stibo/web/std.lang.all/88/51/28928851.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR