2-Chloro-5-methyl-1,4-phenylenediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,5-Diamino-4-chlorotoluene
D2179-25G
TCD2179-025GEA
187.16
USD
TCD2179-025G
2-Chloro-5-methyl-1,4-phenylenediamine ≥98.0% (by GC, titration analysis)
2-Chloro-5-methyl-1,4-phenylenediamine
CAS Number: 5307-03-9
MDL Number: MFCD00190684
Molecular Formula: C7H9ClN2
Molecular Weight: 156.61
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 147
Other Product Names / Synonyms: 2,5-Diamino-4-chlorotoluene
Formula:
C₇H₉ClN₂ MW: 156.61 g/mol Melting Pt: 147 °C |
MDL Number:
MFCD00190684 CAS Number: 5307-03-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...