Di-p-tolylether ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Dimethyldiphenyl Ether

D2178-25G
TCD2178-025GEA 123 USD
TCD2178-025G
Di-p-tolylether ≥98.0%
Di-p-tolylether

CAS Number: 1579-40-4
MDL Number: MFCD00025979
Molecular Formula: C14H14O
Molecular Weight: 198.27
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 285
Melting point (°C): 48
Flash Point (°C): 110


Other Product Names / Synonyms: 4,4'-Dimethyldiphenyl Ether

Formula: C₁₄H₁₄O
MW: 198.27 g/mol
Boiling Pt: 285 °C
Melting Pt: 48 °C
Flash Pt: 110 °C
MDL Number: MFCD00025979
CAS Number: 1579-40-4
https://us.vwr.com/stibo/web/std.lang.all/89/91/28978991.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR