1,3-Diaminopyrene ≥98.0% (by HPLC)

Supplier: TCI America

D2167-100MG
TCD2167-100MGEA 311 USD
TCD2167-100MG
1,3-Diaminopyrene ≥98.0% (by HPLC)
1,3-Diaminopyrene

CAS Number: 92821-64-2
MDL Number: MFCD00142518
Molecular Formula: C16H12N2
Molecular Weight: 232.29
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 184

Formula: C₁₆H₁₂N₂
Melting Pt: 184 °C
MDL Number: MFCD00142518
CAS Number: 92821-64-2
https://us.vwr.com/stibo/web/std.lang.all/89/69/28978969.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR