1,6-Diaminopyrene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
Pyrene-1,6-diamine
D2160-100MG
TCD2160-100MGEA
41.6
USD
TCD2160-100MG
1,6-Diaminopyrene ≥98.0% (by HPLC, total nitrogen)
1,6-Diaminopyrene
CAS Number: 14923-84-3
MDL Number: MFCD00142568
Molecular Formula: C16H12N2
Molecular Weight: 232.29
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 230
Formula:
C₁₆H₁₂N₂ MW: 232.29 g/mol Melting Pt: 230 °C |
MDL Number:
MFCD00142568 CAS Number: 14923-84-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...