Dimethylthiophosphinoyl chloride ≥97.0%

Supplier: TCI America

Synonyms: Chlorodimethylphosphine SulfideDimethylphosphinothioyl Chloride

D2159-1G D2159-5G
TCD2159-001GEA 141.04 USD
TCD2159-001G TCD2159-5G
Dimethylthiophosphinoyl chloride ≥97.0%
Dimethylthiophosphinoyl chloride

CAS Number: 993-12-4
MDL Number: MFCD00014454
Molecular Formula: C2H6ClPS
Molecular Weight: 128.55
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Specific Gravity (20/20): 1.22
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis), Glycosylation, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Chlorodimethylphosphine Sulfide, Dimethylphosphinothioyl Chloride

Formula: C₂H₆ClPS
MW: 128.55 g/mol
Boiling Pt: 78 °C
Density: 1.22 (20 °C)
MDL Number: MFCD00014454
CAS Number: 993-12-4
https://us.vwr.com/stibo/web/std.lang.all/89/56/28978956.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR