TCIAD2148

D2148-25G
TCD2148-025GEA 83.49 USD
TCD2148-025G
TCIAD2148
CAS Number: 81-41-4
MDL Number: MFCD00053082
Molecular Formula: C16H8N4O2
Molecular Weight: 288.27
Purity/Analysis Method: 90.0% (HPLC,N)
Form: Crystal

Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,4-Diamino-9,10-anthraquinone-2,3-dicarbonitrile
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR