N-(2,4-Dinitrophenyl)-L-alanine methyl ester ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Dnp-L-alanine Methyl EsterDnp-Ala-OMe

D2135-1G
TCD2135-001GEA 105 USD
TCD2135-001G
N-(2,4-Dinitrophenyl)-L-alanine methyl ester ≥98.0% (by HPLC)
N-(2,4-Dinitrophenyl)-L-alanine methyl ester

CAS Number: 10420-63-0
MDL Number: MFCD00051742
Molecular Formula: C10H11N3O6
Molecular Weight: 269.21
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 82


Product Categories: Materials Chemistry, Organic Nonlinear Optical (NLO) Materials, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Dnp-L-alanine Methyl Ester, Dnp-Ala-OMe

Formula: C₁₀H₁₁N₃O₆
MW: 269.21 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00051742
CAS Number: 10420-63-0
https://us.vwr.com/stibo/web/std.lang.all/89/13/28978913.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR