3-Decylthiophene ≥98.0%
Supplier: TCI America
D2098-1G
D2098-5G
TCD2098-001GEA
54.77
USD
TCD2098-001G
TCD2098-005G
3-Decylthiophene ≥98.0%
3-Decylthiophene
CAS Number: 65016-55-9
MDL Number: MFCD00143180
Molecular Formula: C14H24S
Molecular Weight: 224.41
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 0.91
Formula:
C₁₄H₂₄S MW: 224.40 g/mol Density: 0.91 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00143180 CAS Number: 65016-55-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...