α-(4-Pyridyl)benzhydryl alcohol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
alpha,alpha-Diphenyl-4-pyridylmethanol
D2089-25G
TCD2089-025GEA
134
USD
TCD2089-025G
α-(4-Pyridyl)benzhydryl alcohol ≥98.0% (by GC, titration analysis)
α-(4-Pyridyl)benzhydryl alcohol
CAS Number: 1620-30-0
MDL Number: MFCD00006434
Molecular Formula: C18H15NO
Molecular Weight: 261.32
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 242
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...