2',4'-Dimethoxyacetoacetanilide ≥95.0% (by total nitrogen basis)

Supplier: TCI America

D2083-25G D2083-500G
TCD2083-025GEA 31.45 USD
TCD2083-025G TCD2083-500G
2',4'-Dimethoxyacetoacetanilide ≥95.0% (by total nitrogen basis)
2',4'-Dimethoxyacetoacetanilide

CAS Number: 16715-79-0
MDL Number: MFCD00043929
Molecular Formula: C12H15NO4
Molecular Weight: 237.26
Purity/Analysis Method: 95.0% (N)
Form: Crystal
Melting point (°C): 96

https://us.vwr.com/stibo/web/std.lang.all/88/68/28978868.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR